Learn More
Agilent BioTek Cell Culture/Petri Dish Adapter for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
Brand:  Agilent BioTek 1222240
 View more versions of this product
Product Code. 15428722
  • 463.00CHF / Each
Only null left
Estimated Shipment: 14-07-2025
Description
For use with Imaging Multi-Mode Readers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Culture/Petri Dish Adapter
Imaging Multi-Mode Reader
Dish diameter: 38.7 mm