Learn More
Biodesign™ Cellulose Dialysis Tubing Roll
Click to view available options
MWCO:
14 kDa
3.5 kDa
8 kDa
Capacity (Metric):
0.32 mL/cm
1.91 mL/cm
19.13 mL/cm
3.57 mL/cm
6.45 mL/cm
Unit Size:
Each
Description
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
- Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
- Just wet in distilled water and use
- Available in 30.5m rolls
- Specially purified for life science research
- Supplied in an airtight, moisture-proof container for maximum shelf life
- Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications
Specifications
Product Type | Dialysis Tubing Roll |
MWCO | 14 kDa |
Material | Cellulose |
Diameter (Metric) | 15.5 mm |
Length (Metric) | 30.5 m |
Capacity (Metric) | 1.91 mL/cm |
Color | Transparent |
Quantity | 1 Each |
For Use With (Application) | For life sciences research |