Learn More
Labconco™ CentriVap™ Complete Concentrator
BIOPHARMA
Comes with everything needed to start processing
Brand:  Labconco™ 7315031
197865.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15142995
  • 203155.00 DKK / Each
Only null left
Estimated Shipment: 03-02-2026
Description
Comes with everything needed to start processing
Uses a combination of centrifugal force, vacuum and heat to speed evaporation.
Includes:
  • Vacuum Concentrator
  • -50°C Cold Trap
  • Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
  • Chemical-resistant diaphragm vacuum pump
  • CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed
Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry
Specifications
Specifications
Centrivap
International
Acid Resistant without Heat Boost
230 V 50 Hz 6 A