Learn More
Unity Lab Services™ Basic Calibration Certificate Multichannel Micropipettor 3 x 10
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand: Unity Lab Services™ umv-p938f
This item is not returnable.
View return policy
Description
- Calibration by gravimetry of the pipettor comprising: verification of delivered volumes according to NF EN ISO 8655 in 10 weighings, three volumes per channel without change of cone
- Production of the COFRAC calibration certificate with declaration of conformity and labeling
- Provision under COFRAC accreditation Nº2-5985
- Calibration, scope available on www.cofrac.fr
- Average time: 1 week
Specifications
Multi-channel Micropipetters | |
2 Weeks | |
Basic Calibration Certificate |
COFRAC N° 2-5985, NF EN ISO 8655 | |
Gravimetric |