Learn More
Pro-Lab Diagnostics™ Nichrome Inoculation Loops
Calibrated to remove a consistent, precise amount of material. Pro-Lab Diagnostics™ Nichrome Inoculation Loops are economical nickel-chrome wire loops in a range of sizes from 1μL to 10μL. The standard and heavy gauge wire loops can be used with insulated holders (sold separately).
52.00€
Product Code | Brand | Diameter (Metric) Inside | Gauge | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Gauge | Price | Quantity & Availability | ||||
11909054
|
Pro-Lab Diagnostics™
PL.104 |
2.26 mm | Standard |
52.00€
Pack of 25 |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||||
11919054
|
Pro-Lab Diagnostics™
PL.105 |
1.59 mm | Standard |
52.00€
Pack of 25 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
||||
11999044
|
Pro-Lab Diagnostics™
PL.102 |
2.91 mm | Standard |
52.00€
Pack of 25 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||||
Description
An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)
- Constructed of economical nickel-chrome in standard or heavy gauge wire
- Available in a range of 6 sizes, 1μL / 1.59mmD to 10μL / 5.05mmD
- Packaged in sets of 25 loops
- Holders with insulated handles sold separately
Specifications
Chrome-Nickel | |
Inoculating Loop |