Learn More
VITLAB™ Class A PMP Graduated Cylinders with Raised Scale
Click to view available options
Graduations:
0.5 mL
1 mL
Unit Size:
Pack of 2
Specifications
Specifications
Class | A |
Graduations | 0.5 mL |
Limit of Error | ±0.25mL |
Shape | Round |
Base Type | Hexagonal |
Height (Metric) | 170 mm |
Type | Graduated Cylinder |