Learn More
VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale feature easy to read, raised, embossed blue scale and ring marks at the primary scale points.
Brand:  VITLAB™ 651081
 View more versions of this product
Product Code. 10412321
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Highly transparent polypropylene graduated cylinder
  • Hexagonal base with bottom studs provides high stability
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
  • To preserve markings, do not clean at temperatures exceeding 60°C
Specifications
Specifications
Polypropylene
DIN 12681, ISO 6706
B
5 mL
Round
Blue
360 mm
±5 mL
6Pack
58 mm
±5mL
Food Industry and Volume Measurement
Hexagonal
Graduated Cylinder