Learn More
Cleaning and Hygiene Sup Luer-Tip Syringe
Product Code. 12399199
Click to view available options
Unit Size:
Pack of 100
Cleaning and Hygiene Sup Luer-Tip Syringe for research use only, unless otherwise indicated.
  • For gases or liquids
  • Individually wrapped in sealed envelope
  • With polypropylene barrel and plunger, and rubber piston with double contact ring
  • Sterilised by ethylene oxide
  • Without needle
Specifications
Certifications/Compliance ISO 7886 (DIS 7886)
Material Plastic
For Use With (Application) For gases or liquids
Product Type Luer-tip Syringe
Disposable Single-use
Graduations 0.2 mL
Quantity 100/Pk.
Volume (Metric) 5 mL
Connection Type Luer Tip
Graduated Yes