Learn More
Memmert™ Clip for 300 mL Flask
Clip for 300 mL flask (14 flasks max.)
Brand:  Memmert™ WTB35B02694
216.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17180365
  • 226.00 DKK / Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Accessory
  • Max. flasks see sketch on demand
Specifications
Specifications
WTB35
Clip for 300 mL flask