missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Chest Freezers, 75 mm Box Height, with Boxes
Comfort Rack suitable for use with all chest freezer models.
Brand: Tenak™ TE24536B
This item is not returnable.
View return policy
Description
- Racks with spring locks mounded on each shelf to secure the cryoboxes
- Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
- Measure the inside height of your freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Spring Locks | |
Stainless Steel | |
Chest Frseezers | |
75 mm (Box) |
Boxes | |
5 | |
140 x 140 x 406 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction