Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 45mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
Brand: Tenak™ TE35546
2264.55 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Racks can either be used as one plate per shelf or the plates can be stacked
- With an alert preventing the shelves from sliding out
- External footprint is 135 x 92mm
Specifications
Microtiter Plate Rack | |
Plates (36) | |
Yes | |
1 Pack | |
135 mm | |
45 mm |
6 x 6 | |
36 | |
Microtiter Plates, Upright Freezers | |
540 x 135 x 279 mm | |
279 mm | |
540 mm |