Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 65mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
Brand:  Tenak™ TE35604
1972.23 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12407110
  • 2025.00 DKK / Each
Only null left
Estimated Shipment: 15-09-2025
Description
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • With an alert preventing the shelves from sliding out
  • External footprint is 135 x 92mm
Specifications
Specifications
Microtiter Plate Rack
Plates (28)
Yes
1 Pack
135 mm
65 mm
4 x 7
28
Microtiter Plates, Upright Freezers
630 x 135 x 266 mm
266 mm
630 mm