Learn More
Buerkle™ Compression-Type Fitting for Buerkle™ Fish-Clip
Buerkle™ Compression-Type Fitting for Buerkle™ Fish-Clip allows you to easily fix the fish-clip on differnt bottle threads.
61.73 DKK - 230.00 DKK
Specifications
Description | Equipment for fish-clip |
---|---|
Material | PVDF |
For Use With (Equipment) | Fish-Clip™ Cell Cultures Flasks |
Product Type | Compression-type fitting |
Autoclavable | Autoclavable |
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | ||||
10487572
|
Buerkle™
8280-0256 |
25 mm | 19/26 NS-cone | N/A | |||||
10271542
|
Buerkle™
8280-0321 |
32 mm | 29/32 NS-cone | N/A | |||||
10561262
|
Buerkle™
8280-0255 |
32 mm | Without NS-cone | N/A | |||||
10500892
|
Buerkle™
8280-0254 |
25 mm | Without NS-cone | N/A | |||||
Description
- PVDF
- Autoclavable
- Good chemical resistance
- Can fix the fish-clip easily on differnt bottle threads
Specifications
Equipment for fish-clip | |
Fish-Clip™ Cell Cultures Flasks | |
Autoclavable |
PVDF | |
Compression-type fitting |