Learn More
Fisherbrand™ Conductivity Sensor
The machine can be equipped with conductivity control.
Brand:  Fisherbrand™ 2908130
Product Code. 17107784
  • 16260.00 DKK / Each
Only null left
Estimated Shipment: 26-01-2026
Description
  • Conductivity for a final rinse can be set and the number of attempts the machine must try to reach the set value
  • Conductivity control is easily set in the programming of the rinse phase
Specifications
Specifications
Conductivity Sensor