Learn More
WTW™ Connection Cable with BNC Plug for WTW™ pH/ORP Electrode
Brand: WTW™ 108114
395.43 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
1 m Length | |
Connection Cable | |
BNC plug, socket S7 | |
1 m |