missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated NSUN2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66580-20UL
This item is not returnable.
View return policy
Description
The NSUN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NSUN2. This antibody recognizes human antigen. The NSUN2 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
NSUN2 | |
Monoclonal | |
1000 μg/mL | |
FLJ20303; MISU; SAKI; TRM4 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human | |
100 kDa | |
Liquid |
Immunocytochemistry/Immunofluorescence | |
2H5A2 | |
CoraLite Plus 488 | |
Mouse | |
Protein A purification | |
RUO | |
54888 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction