missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated PPAR Gamma Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60127-100UL
This item is not returnable.
View return policy
Description
CoraLite™ Plus 488-conjugated PPAR Gamma Mouse Monoclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
PPAR Gamma | |
Monoclonal | |
1000 μg/mL | |
CIMT1; NR1C3; PPARG1; PPARG2; PPARgamma | |
Recombinant Protein | |
100 μL | |
Primary | |
Human | |
Conjugated Antibody | |
25 |
Not Available | |
4E12F10 | |
CoraLite Plus 488 | |
Mouse | |
Protein G purification | |
RUO | |
5468 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction