Learn More
Corning™ Bar Coded Storage Tubes:2D Bar Coded
Shop All Corning Products
Click to view available options
Includes:
Rack
Packaging:
Rack
Product Type:
Barcoded Latch Rack
Round Tube
Unit Size:
Each
Pack of 384
Description
Feature a synchronized coding scheme that minimizes confusion and the risk of duplication, while enabling safe and secure sample identification.
- Maximum Identification — Synchronized 2D and linear bar code
- Maximum Information — 14 x 14 dot 2D bar code
- Bar Code Stability — Laser-etched, not an attached label
- Compound Compatibility — Polypropylene construction throughout creates inert vessel
- Temp. tolerance: -180 to +110°C (screw cap); -80 to +110°C (TPE cap)
- Length: 14.0mm
- 384/rack
Specifications
Specifications
Product Type | Round Tube |
Autoclavable | Autoclavable |
Capacity (Metric) | 120 μL |
Material | Polypropylene |
For Use With (Application) | Enhanced sample storage, identification and retrieval |
Includes | Rack |
Length (Metric) | 14 mm |
Packaging | Rack |
Shape | Round |
Well Shape | Round |