Learn More
Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile
Allows simultaneous processing
1280.55 DKK - 1317.66 DKK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10631272
![]() |
Corning™
3520 |
15 mm |
1317.66 DKK
Pack of 8 |
In Stock
Log in to see stock. |
|||||
10515531
![]() |
Corning™
3521 |
24 mm |
1280.55 DKK
Pack of 8 |
Estimated Shipment: 17-12-2025
Log in to see stock. |
|||||
Description
Specifications
Carrier Kit | |
8/Pk. | |
8/Cs. |