Learn More
Marienfeld Superior™ Counting Chamber with Dark Line Double Net Ruling
Marienfeld Superior™ Counting Chamber with double net ruling are precise measuring instruments for investigation of cell suspensions
80.00 € - 90.00 €
Specifications
Product Type | Counting chamber, Dark line |
---|---|
Grid Type | Burker |
Description | CE-marked (IVD) |
Cell Size | 0.0025 mm2 |
Material | Special optical glass |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15817045
|
Marienfeld Superior™
0640211 |
Two cover glasses 20 x 26 mm and two clamps |
90.00 €
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||||
11314052
|
Marienfeld Superior™
06 402 10 |
Two cover glasses 20 x 26 mm |
80.00 €
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
Description
- Precise measuring instrument for determination of the number of particles in liquids
- Made of special optical glass
- Equipped with two grids
- With 2 haemacytometer cover glasses 20x26 mm
- Individually packed in a transparent plastic box, 10 boxes in a carton
Specifications
Counting chamber, Dark line | |
CE-marked (IVD) | |
Special optical glass | |
0.1 mm |
Burker | |
0.0025 mm2 | |
1 |