Learn More
Tenak™ White Cryobox with Dividers, 130 mm
Cryobox with dividers for cryogenic storage.
Brand: Tenak™ TE22245
1754.82 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Cryobox
- For use with cryogenic storage applications
Specifications
White | |
9 x 9 Dividers |