Learn More
SI Analytics™ Data Cable
Data cable, 1.5 m length for use with TitroLine™ titrators and Mettler balances
Brand: SI Analytics™ 285223594
507.35 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 1.5 m length
- For TitroLine™ 6000,7000, 7750, 7800, 7500 KF, 7500 KF trace or Mettler AB-S, PG series balances
Specifications
Data cable 1.5 m | |
1.5 m | |
TitroLine 6000,7000, 7750, 7800, 7500 KF, 7500 KF Trace or Mettler AB-S, PG Series Balances |