Learn More
BD DIFF-SAFE™ Blood Dispenser
Click to view available options
Unit Size:
Pack of 1000
Description
The BD Vacutainer™ system is a closed system optimally designed for controlling pre-analytical variability throughout the specimen collection and handling process. The DIFF-SAFE™ blood dispenser prepares blood slides from a blood collection tube without removing the stopper from the tube.
- Features a non-sterile cannula that inserts easily through the rubber stopper of a blood collection tube
- Invert the tube and press the dispenser against a slide to dispense a drop to prepare a blood smear
- Compatible with BD Vacutainer™ tubes with Hemogard™ closures
- Limits exposure to blood samples, which may contain bloodborn pathogens
- Keeps closure in place to prevent spills, leaks and contamination of samples
Specifications
Specifications
Color | White |
For Use With (Application) | Preparing blood smear slides from a blood collection tube. |
Disposable | Single-use |
For Use With (Equipment) | Vacutainer Blood Collection Tubes with Hemogard Closures |
Connection Type | Cannula |
Sterility | Non-sterile |