Learn More
Thermo Scientific™ Dionex™ IonPac™ AS17-C IC Columns
Quickly determine common inorganic anions in high-purity water matrices.
Marca: Thermo Scientific™ 66294
Descrizione
Quickly determine common inorganic anions in high-purity water matrices with the Thermo Scientific™ Dionex IonPac AS17-C column. This low-capacity column is optimized for fast gradient separation of inorganic anions in simple sample matrices. It is also ideal for fast gradient separation of sulfur species, including sulfite, sulfate, and thiosulfate. The Dionex IonPac AS17C column is the recommended replacement for all Dionex IonPac AS17 column anion applications.
Make Gradient Separations as Easy as Isocratic RunsThe Dionex IonPac AS17-C column is designed to be used in conjunction with the Dionex Eluent Generator. The Dionex Eluent Generator automatically produces potassium hydroxide eluents from water, making gradient separations as easy as isocratic runs. Using the Dionex Eluent Generator under gradient conditions, common inorganic anions can easily be separated in a variety of sample matrices including high purity water, drinking water, wastewater, process streams, and scrubber solutions.
Achieve Superior Chromatographic Performance- Fast gradient separation of inorganic anions in high purity water matrices
- Easy suppression of hydroxide eluent to a very low background with the Thermo Scientific™ Dionex™ Anion Self-Regenerating Suppressor (ASRS™ 300) for trace level determinations (including fluoride) even with a large loop injection
- Superior retention and quantification of fluoride, acetate, propionate, and formate
- Meets performance requirements specified in U.S. EPA Method 300.0 (A)
- Operates at ambient or elevated temperatures
- Column selectivity is optimized for operating temperature of 30°C to ensure reproducible results
- Compatible with Reagent-Free™ Ion Chromatography (RFIC™) systems
- Compatible with organic solvents to enhance analyte solubility, modify column selectivity, or for effective column clean-up
Specifica
30 μeq | |
Analytical Column | |
U.S. EPA Method 300.0 (A) | |
Hydroxide | |
Anion Exchange | |
Low | |
4000 psi (275 bar) | |
10.5 μm | |
Standard Bore | |
Each 1 | |
75 nm | |
Responsibly packaged |
PEEK | |
55% DVB | |
30 μeq | |
1.0 to 3.0 mL/min. | |
ICS-2000, ICS-3000 IC System | |
Alkanol Quaternary Ammonium Ion | |
pH 0 to 14, 0 to 100% HPLC Solvents | |
IonPac AS17-C | |
Ethylvinylbenzene/Divinylbenzene | |
250 mm | |
4 mm |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.