Learn More
BRAND™ Dosing Element for Dispensette™ S Trace Analysis Bottletop Dispensers
Replacement dosing element for all models of Dispensette™ S Trace Analysis dispensers, 1-10 mL. Includes certificate of performance.
Brand: BRAND™ 708035
2887.02 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement dosing element for BRAND™ Dispensette™ S Trace Analysis 1 to 10 mL
- Fits instruments with tantalum or platinum-iridium spring
- Includes certificate of performance
Specifications
10 mL | |
Dispensette S Trace Analysis | |
Quality Certificate |