Learn More
Sampling Systems™ Double Bagged - USP Class VI PharmaScoops™
Double Bagged PharmaScoops™ are made from high grade medical USP Class VI HDPE. They are available in a range of sizes from 125 mL to 1000 mL. This premium range is double bagged. A box liner is also used giving a third layer of protection.
938.00 DKK - 1805.87 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17544696
![]() |
Sampling Systems™
9045H-125S |
125 mL | N/A | ||||||
30286578
![]() |
Sampling Systems™
9045H-125S |
125 mL |
1805.87 DKK
Pack of 40 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17554696
![]() |
Sampling Systems™
9045H-250S |
250 mL |
1070.00 DKK
Pack of 25 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
17534696
![]() |
Sampling Systems™
9045H-1000S |
1000 mL |
938.00 DKK
Pack of 10 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
Description
- These sterile scoops are commonly used in the manufacture and sampling of cosmetics, food and pharmaceuticals.
- These scoops all have an ergonomic handle which is positioned high up the body so as to keep the hand as far away from the powder as possible.
- The scoops are designed to be used straight from the bag.
- The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
- Made from food grade, FDA and EC compliant HDPE.
- HDPE has good chemical resistance and can be easily recycled.
- The funnels are sterilised by gamma irradiation.
- USP Class VI - High quality medical grade polymer that has also been independently tested to USP 661.
- The scoops are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
- Each scoop is 100% inspected before packing.
Specifications
HDPE | |
Sterile | |
Double Bagged |