Learn More
DWK Life Sciences DURAN™ Connection Piece, with standard ground cone, 90° angle
The connector allows the connection of equipment via a hose.
153.00 DKK - 198.00 DKK
Specifications
Quantity 1 Ea.
View More Specs
Products 3
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
10767301
View Documents
DWK Life Sciences
243000604
NS 14/23
153.00 DKK
Each
Only null left
In Stock
Add to basket
10602842
View Documents Promotion Details
DWK Life Sciences
243000904
NS 29/32
198.00 DKK
Each
Only null left
Estimated Shipment: 22-07-2025
Add to basket
10203701
View Documents
DWK Life Sciences
243000801
NS 24/29
156.00 DKK
Each
Only null left
In Stock
Add to basket
Description
Description
Connectors are transition pieces with olive and have an olive instead of the second standard ground joint. Tubes can be connected via this olive, which allow e.g. the connection of a vacuum pump.
  • Tight closure due to standard ground joint
  • 90° angle
  • With standard ground cone
  • Easy and tight connection of hoses
  • Fired-on, highly durable white print
Specifications
Specifications
1 Ea.