Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
10.00 EUR - 23.00 EUR
Product Code | Brand | Cone Size | Standard Taper Joint No | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No | Price | Quantity & Availability | ||||
10088450
|
Lenz Laborglasinstrumente™
01311414 |
NS 14/23 | GL 14 |
10.00 EUR
Each |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||||
10597741
|
Lenz Laborglasinstrumente™
01311419 |
NS 19/26 | GL 14 |
12.00 EUR
Each |
Estimated Shipment: 30-09-2025
Log in to see stock. |
||||
10692411
|
Lenz Laborglasinstrumente™
01311819 |
NS 19/26 | GL 18 |
17.00 EUR
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||
10324001
|
Lenz Laborglasinstrumente™
01312519 |
NS 19/26 | GL 25 |
20.00 EUR
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
||||
10363901
|
Lenz Laborglasinstrumente™
01311429 |
NS 29/31 | GL 14 |
22.00 EUR
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||
10708931
|
Lenz Laborglasinstrumente™
01311424 |
NS 24/29 | GL 14 |
22.00 EUR
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||
10160121
|
Lenz Laborglasinstrumente™
01312529 |
NS 29/32 | GL 25 |
23.00 EUR
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||
10405801
|
Lenz Laborglasinstrumente™
01311824 |
NS 24/29 | GL 18 |
21.00 EUR
Each |
Estimated Shipment: 13-10-2025
Log in to see stock. |
||||
10632421
|
Lenz Laborglasinstrumente™
01311829 |
NS 29/32 | GL 18 |
19.00 EUR
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||
10002944
|
Lenz Laborglasinstrumente™
01313229 |
NS 29/32 | GL 32 |
18.00 EUR
Each |
Estimated Shipment: 24-10-2025
Log in to see stock. |
||||
Description
- Acc. to DIN 12257
- Available in a range of socket and thread sizes
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
1 Ea. | |
Duran Glass |