Learn More
Lenz Laborglasinstrumente™ Tubes with Socket and Cone
Provides for quick and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Tubes with Socket and Cone are manufactured from durable DURAN™ borosilicate glass.
121.00 DKK - 148.00 DKK
Specifications
Quantity 1Pack
View More Specs
Products 3
Product Code Brand Cone Size Price Quantity & Availability  
Product Code Brand Cone Size Price Quantity & Availability  
10589951
View Documents
Lenz Laborglasinstrumente™
01101411
Socket Size/Cone Size: NS 14/23
121.00 DKK
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
10671892
View Documents
Lenz Laborglasinstrumente™
01101422
Socket Size/Cone Size: NS 19/26
129.00 DKK
Each
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11846144
View Documents
Lenz Laborglasinstrumente™
01101444
Socket Size/Cone Size: NS 29/32
148.00 DKK
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Description
Description
  • Available in a range of socket/cone sizes
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featurig high taper accuracy and excellent tightness
Specifications
Specifications
1Pack