Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable, with standard ground cone, HP, HQ
KPG bearing sleeve is a precisely fitting holder for guiding the KPG agitator shaft.
608.00 DKK - 635.00 DKK
Specifications
Quantity 1 Ea.
Joint Size 10 mm
View More Specs
Products 2
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
10303561
View Documents
DWK Life Sciences
245285603
HQ 10, NS 29/32
635.00 DKK
Each
Only null left
In Stock
Add to basket
10008230
View Documents
DWK Life Sciences
245235504
HP 10, NS 24/29
608.00 DKK
Each
Only null left
Estimated Shipment: 22-07-2025
Add to basket
Description
Description
KPG agitators consist of a KPG agitator shaft and a KPG bearing sleeve. KPG bearing parts are manufactured with high accuracy and uniformity. The specified tolerances for nominal diameter and fit are maintained by careful checks. This guarantees the interchangeability of the KPG bearing sleeves and KPG stirrer shafts. Due to the low bearing clearance between the running surfaces, almost absolute sealing and completely quiet running can be achieved with suitable lubrication. KPG agitators are an alternative to magnetic stirrers and are used for reaction vessels that are not accessible from below when very viscous reaction mixtures are to be stirred or a high stirring speed is required.
  • High accuracy and uniformity within defined tolerances for nominal diameter and fit
  • Tight closure due to standard ground joint
  • Ground and polished bearing surface
  • Interchangeable
Specifications
Specifications
1 Ea.
10 mm