Learn More
DWK Life Sciences DURAN™ PURE Premium Screw Cap
High-quality closures developed for the needs of the pharmaceutical and biotech industry
315.00 DKK - 623.00 DKK
Specifications
Color | Clear |
---|---|
For Use With (Application) | Manufacturing of pharmaceutical drugs and biotech products; Storage and transport of APIs, bulk drug intermediates, ingredients and media |
Material | PFA, PTFE, Silicone |
Quantity | 5/Pk. |
Autoclavable | Autoclavable |
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | |||||
11679808
![]() |
DWK Life Sciences
891088679 |
GL 45 |
623.00 DKK
Pack of 5 |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||||
12866573
![]() |
DWK Life Sciences
891081306 |
GL 25 |
315.00 DKK
Pack of 5 |
In Stock
Log in to see stock. |
|||||
Description
High-quality plastic screw caps with GL 25/GL 45 thread sizes, developed for the needs of the pharmaceutical industry. DURAN™ PURE Premium Screw Caps are manufactured in clean rooms and made from PFA with highly inert PTFE coated silicone seal. Designed for use with DURAN™ PURE GL 25/ GL 45 Bottles. The screw caps are compatible with autoclaving and other common sterilization methods. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. DURAN™ PURE Premium Screw Caps are available with the documentation required by regulatory authorities and meet the requirements of the international pharmacopoeias.
- High cleanliness due to manufacturing in clean rooms
- Single batch delivery
- Certificate of Analysis (CoA)
- Visual and dimensional inspection
- Change control management and information of customer in case of changes
- 100% traceability of all manufacturing steps
- Extractables report available
- Drug Master File
- USP <88> Class VI Pharmaceutical grade materials
Specifications
Clear | |
PFA, PTFE, Silicone | |
Autoclavable |
Manufacturing of pharmaceutical drugs and biotech products; Storage and transport of APIs, bulk drug intermediates, ingredients and media | |
5/Pk. |