missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt dihydrate, BAKER ANALYZED™, J.T. Baker™

Product Code. p-7220829
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym Ethylenediaminetetra-acetic acid disodium salt dihydrate , Edetate disodium dihydrate , Ethylenediaminetetraacetic acid, disodium salt, dihydrate , Kestranal™ 2 S 2 H2O , Ethylenedinitrilotetra-acetic acid disodium salt dihydrate , Titriplex III , Kestranal™ 2 S dihydrate , Edetic acid disodium salt dihydrate , Disodium dihydrogen ethylenediaminetetraacetate dihydrate
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 255°C
pH 4.0 - 6.0 (5% Solution at 25°C)
Assay Percent Range 99.0 to 101.0%
Packaging Plastic bottle for solids
Quantity 250 g
Insoluble Matter max. 0.005%
compliance-icons compliance-icons
Product Identifier
  • EDTA disodium salt dihydrate BAKER ANALYZED Reagent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : EDTA disodium salt dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.