missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF5A2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
968.42 DKK - 2619.89 DKK
Specifications
Antigen | EIF5A2 |
---|---|
Clone | 1F9B2 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30237031
|
Proteintech
67907-1-IG-150UL |
150 μL |
2619.89 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30236421
|
Proteintech
67907-1-IG-20UL |
20 μL |
968.42 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EIF5A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF5A2. This antibody recognizes human, rat antigen. The EIF5A2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
EIF5A2 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
56648 | |
Recombinant Protein | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
1F9B2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
Unconjugated | |
Mouse | |
Human, Rat | |
EIF-5A2; eIF5AII | |
IgG1 | |
Protein G purification | |
Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title