Learn More
Eppendorf™ Centrifuge Bottles
Wide mouth plastic centrifuge bottles.
1616.44 DKK
Specifications
Includes | Screw cap |
---|---|
Autoclavable | Autoclavable |
Max. Force | 3250 x g |
Mouth | Wide |
For Use With (Equipment) | 1 x 400mL Adapter for super capacity 4 x 400mL rotor A-4-81 |
Description
For use with Eppendorf Centrifuges 5804/5804R, 5810/5810R.
Specifications
Screw cap | |
3250 x g | |
1 x 400mL Adapter for super capacity 4 x 400mL rotor A-4-81 | |
Screw Cap |
Autoclavable | |
Wide | |
Polypropylene |