missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute, Solveco
Click to view available options
Quantity:
3 x 5 L
12 x 1 L
Description
Well suited for routine laboratory and analysis. Solveco Ethanol Absolute is fermented and rectified ethanol with very high purity. This PA quality solution has a natural origin.
Specifications
Specifications
Absorbance | ≤0.500 (UV 5 cm, 240 nm) |
Color | Colorless |
Density | ≤0.790 g/cm3 |
Boiling Point | 78°C |
Quantity | 3 x 5 L |
Packaging | HDPE Drum |
Formula Weight | 46.07g/mol |
Percent Purity | 99.9% |
Grade | AG |
Chemical Name or Material | Ethyl Alcohol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction