Learn More
Sartorius™ External Adjusting Weight Without DKD Calibration Certificate
Test weight weight set for special weighing instruments
Brand: Sartorius™ YSS43-00
1447.43 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 5 g, 50 g and 5 kg in wooden case, 30 g in a plastic container
- Calibration weight 30 g ± 0.3mg
- DAkkS Certificate
- Special, non-magnetizable steel
Specifications
DAkkS Certificate | |
Calibration weights | |
Special Steel, Non-magnetizable | |
5 g, 50 g, 5 kg, 30 g calibration weight |