Learn More
Ferroin, Chem Lab™
Brand: Chem Lab CL07.0609.0100
Quantity | 100 mL |
---|
This item is not returnable.
View return policy
Specifications
0°C | |
Odorless | |
4 to 7 | |
Liquid |
Soluble in water | |
Red | |
100 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ferroin
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : iron sulphide