Learn More
Fisher Scientific™ Bath Circulator Accessories: Plumbing Packages
Plumbing kits for use with bath circulators and chillers
Brand: Fisher Scientific™ 1600147
2506.88 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Plumbing kits for use with bath circulators and chillers
- Plumbing kit
- Material: Viton
Specifications
Isotemp™ circulating bath | |
-25°C to +100°C | |
Plumbing Kit, Insulated | |
Viton |