Learn More
Fisherbrand™ Model 705 Sonic Dismembrator Accessory
4 Tip horn for use with 500 or 700 watt sonicator systems; replacement tip also available.
3595.00 DKK - 14995.00 DKK
Requires: Cat. No. FB4659
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
15320382
|
Fisherbrand™
FB4660 |
Replacement Tip | 4-Tip Horn ( #4659) |
3595.00 DKK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
||||
15310382
|
Fisherbrand™
FB4659 |
4-tip Horn | FB705 Sonic Dismembrator and Q700 Sonicator |
14995.00 DKK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
||||
Description
- For processing 0.5 to 15ml volumes
- Process 4 samples at one time