Learn More
Fisherbrand™ Wash Bottles with Adjustable Sprayer
Handle hot/cold, aqueous/solvent-based solutions
238.00 DKK - 277.00 DKK
Specifications
Color | White |
---|---|
Includes | Polypropylene adjustable sprayer |
Material | HDPE |
Sterility | Non-sterile |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11714356
|
Fisherbrand™
90010-81 |
240 mL |
277.00 DKK
Pack of 3 |
Estimated Shipment: 20-10-2025
Log in to see stock. |
|||||
11795873
|
Fisherbrand™
90023-74 |
1000 mL |
238.00 DKK
Each |
Estimated Shipment: 13-11-2025
Log in to see stock. |
|||||
Description
Handle hot/cold, aqueous/solvent-based solutions using Fisherbrand™ Wash Bottles with Adjustable Sprayer.
Fully adjustable spray pattern: 'fine mist' for TLC plates; 'needle' for rinsing glassware; 'big stream' for powerful jet needed to wash filters.
- Bottle is made of high-density polyethylene
- Sprayer is polypropylene
- Trigger allows one-handed, controlled delivery with light pressure
- Engraved markings show how much fluid is left
- Squat shape takes up little bench space and ensures a low center of gravity
Specifications
White | |
HDPE | |
Yes | |
Reusable | |
Threaded | |
10.16 cm | |
4 in. | |
Threaded |
Polypropylene adjustable sprayer | |
Non-sterile | |
Not Autoclavable | |
No | |
Squat | |
Spray Bottle | |
Polypropylene |