Learn More
Cole-Parmer™ Stuart™ Florentine Flask, Plastic-coated For Stuart™ Rotary Evaporators
Plastic coating prevents shattering. Used with Stuart™ Rotary Evaporators.
Brand: Cole-Parmer™ 10096-59
3497.92 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Stuart™ rotary evaporators
- Plastic coated glass
- 29/32 socket
Specifications
Evaporating flask | |
Plastic-coated Glass | |
Flat |
2 L | |
29/32 |