Learn More
Rogo Sampaic™ Forceps
94.00€ - 102.00€
Specifications
Product Type | Forceps |
---|---|
Material | Stainless Steel |
For Use With (Application) | For Tissue |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11718234
|
Rogo Sampaic™
FMJ008 |
12 cm |
102.00€
Pack of 10 |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||||
11748234
|
Rogo Sampaic™
FMJ013 |
14 cm |
94.00€
Pack of 10 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
Specifications
Forceps | |
Stainless Steel | |
For Tissue |