Learn More
Invitrogen™ FTO Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551074
This item is not returnable.
View return policy
Description
FTO Polyclonal Antibody detects endogenous levels of FTO protein.
FTO is a novel protein and a member of the non-heme dioxygenase (Fe(II)-and 2-oxoglutarate-dependent dioxygenases) superfamily. Amino acid conservation patterns support this hypothesis and indicates that both 2-oxoglutarate and iron should be important for FTO function. Though not much is yet known about FTO, it is thought that the protein may somehow modify the activity of genes involved in metabolism and fat storage, which in turn may influence a person's risk of obesity.
Specifications
FTO | |
Polyclonal | |
Unconjugated | |
Fto | |
AlkB homolog 9; ALKBH9; alpha-ketoglutarate-dependent dioxygenase FTO; AW743446; BMIQ14; fat mass and obesity associated; fat mass and obesity-associated protein; fatso; FTO; GDFD; KIAA1752; m6A(m)-demethylase FTO; MGC5149; mKIAA1752; mRNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; mRNA N(6)-methyladenosine demethylase FTO; Protein fatso; protein fto; RGD1305121; tRNA N1-methyl adenine demethylase FTO; U6 small nuclear RNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; U6 small nuclear RNA N(6)-methyladenosine-demethylase FTO | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26383, 291905, 79068 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q2A121, Q8BGW1, Q9C0B1 | |
Fto | |
Synthesized peptide derived from the N-terminal region of human FTO. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |