Learn More
Ekastu Safety™ Gastight Goggle
Click to view available options
UV Protection:
Yes
Description
- DIN EN 166 1 5 – SN
- Full view goggles resistant to chemicals
- Gas tight and dustproof
- Made of rubber which is kind to the skin
- With clear three-layer sandwich lenses
- Anti-fogging
- Large field of vision
- Quick and easy lens replacement
- Comfortable, pressure-free fit
- 99.99% UV protection with innovative UV 380 technology
Specifications
Specifications
Certifications/Compliance | DIN EN 166 1 5 – SN |
Lens Coating | Anti-fog |
Product Type | Gastight Goggle |
For Use With (Application) | Gas and dust applications, chemical resistant |
Frame Color | Black |
No. per Pack | 1 Ea. |
Material | Rubber |
UV Protection | Yes |