Learn More
Macherey-Nagel™ Glass Fiber Wadding
Product Code. 10209841 Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
Used GC Liners, Packed GC Columns
Unit Size:
25g