Learn More
BUCHI Glass Woulff Flask
Brand: BUCHI 025519
3632.32 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Glass | |
Plastic | |
800 mL |
Woulff Flask | |
1 Each |