Learn More
Equip Labo™ Gooseneck Tap
149.00€ - 298.00€
Specifications
Type Gooseneck Tap
Quantity 1 Ea.
View More Specs
Products 3
Product Code Brand Description Height (Metric) Price Quantity & Availability  
Product Code Brand Description Height (Metric) Price Quantity & Availability  
12608832
View accessory products View Documents
Equip Labo™
4768
Backsplash 250 mm
190.00€
Each
Only null left
Estimated Shipment: 01-08-2025
Add to basket
12519225
View accessory products View Documents
Equip Labo™
4778
Surface Mount 250 mm
149.00€
Each
12596616
View accessory products View Documents
Equip Labo™
4790
Remote Control 150 mm
298.00€
Each
Specifications
Gooseneck Tap
1 Ea.