Learn More
Grace™ Reveleris™ Silica Flash Cartridges
Ideal for speed, resolution and capacity
POA - 144.84 €
Specifications
Material | Silica |
---|---|
Type | Reveleris SRC Silica Flash Cartridge |
For Use With (Application) | For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems |
Product Code | Brand | Bed Weight (Metric) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight (Metric) | Type | Price | Quantity & Availability | ||||
12985911
|
Grace™
145146130 |
4 g | Reveleris SRC Silica Flash Cartridge | N/A | N/A | N/A | |||
10216154
![]() |
Grace™
145146132 |
40 g | Reveleris SRC Silica Flash Cartridge |
POA
Each |
N/A | ||||
10597534
![]() |
Grace™
145146134 |
120 g | Reveleris SRC Silica Flash Cartridge |
POA
Each |
N/A | ||||
Description
Silica flash cartridges.
- Withstand higher pressures (up to 200psig) and deliver greater loading capacity
- Superior, high load silica for excellent resolution
- Optimally packed to give high plate count
- Tighter particle size distribution and minimal fines keep backpressure low
- RFID tag streamlines Reveleris system (sold separately) operation by auto-loading cartridge specifications
- Compatible with other popular flash systems
Specifications
Silica | |
Reveleris SRC Silica Flash Cartridge | |
For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems |