Learn More
Buerkle™ Graduated Polypropylene Cylinder with Embossed Blue Gradations
Measuring cylinder, tall shape, according to DIN 12 680(Glass)/ISO 6706, with hexagonal base. For use in laboratories, chemists, in workshops, during outdoor field work and wherever volumetric measurements have to be made during production.
60.93 DKK - 399.12 DKK
Specifications
Material | Polypropylene |
---|---|
Certifications/Compliance | Category B DIN 12681/ISO 6706 |
Scale | Embossed |
Includes | Embossed blue graduation with rings at the main points |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
10674723
|
Buerkle™
7503-0050 |
50 mL | 1 mL |
60.93 DKK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
17290674
|
Buerkle™
7503-0500 |
500 mL |
184.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
10347192
|
Buerkle™
7503-0100 |
100 mL | 1 mL |
88.85 DKK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||
10738954
|
Buerkle™
7503-0250 |
250 mL | 2 mL |
123.47 DKK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
10245882
|
Buerkle™
7503-1000 |
10 mL |
222.20 DKK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10024612
|
Buerkle™
7503-2000 |
2000 mL | 20 mL |
399.12 DKK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
Description
- PP, highly transparent
- Category B (DIN 12681 / ISO 6706)
- Embossed blue graduation with rings at the main points
- Shatterproof
- Adjusted to “In”
Specifications
Polypropylene | |
Embossed | |
Autoclavable | |
Blue | |
Graduated Cylinder |
Category B DIN 12681/ISO 6706 | |
Embossed blue graduation with rings at the main points | |
Round | |
Hexagonal |