Learn More
Sampling Systems™ Graduated SteriWare Beakers
This range of graduated beakers are crucial in the production of drugs, chemicals and cosmetics. Individually bagged to prevent cross contamination, these beakers also have a pouring spout to ensure the convenient handling of liquids.
Brand: Sampling Systems™ 8008P-100
454.36 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made from food grade, FDA and EC compliant polypropylene
- Available sterile and non-sterile
- Wide range of sizes
- 100% manufactured and packed in a cleanroom
Specifications
100 mL | |
Non-sterile | |
Single-use | |
FDA | |
Low Form |
Polypropylene | |
Autoclavable | |
Yes | |
Molded Scale |